Learn More
Electron Microscopy Sciences Kodak Rapid Fixer, 1 Gal Kit
SDP
Catalog No. 5026811
Encompass_Preferred
A rapid-acting hardening fixer for automated and tray processing. Complete fixer. Kit makes 1 gal: 32oz solution A and 3.6 oz solution B.
Catalog No. 50-268-11 Supplier Electron Microscopy Sciences Supplier No. 74313
A rapid-acting hardening fixer for automated and tray processing. Complete fixer. Kit makes 1 gal: 32oz solution A and 3.6 oz solution B.