Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Kodiak™ Jersey-Lined Premium Cow Leather Welding Gloves

Catalog No. 19055403 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Coating Material:
Uncoated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coating Material
19-055403 Uncoated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-055403 Supplier MCR Safety Supplier No. 4620
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kodiak™ Welder's Gloves are constructed of premium quality cow side leather with Aramid Fiber stitching for durability and burn resistance.

  • Premium quality cow side leather
  • Cotton full sock jersey lining
  • Sewn with Aramid Fiber for durability
  • Self hemmed cuffs

Specifications

Coating Material Uncoated
Color Yellow
Grade Premium
Material Side Split Cowhide Leather
Size X-Large
Style Gunn
For Use With (Application) Welding
Glove Cuff Style Gauntlet
Liner Material Jersey
Series Kodiak™ 4620
Thumb Style Wing
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.