Learn More
Med Vet International Koe Concentrate 16oz
Catalog No. 502831081
Encompass_Preferred
Eliminates, does not mask, even old, impregnated odors from cages, runs, walls and other large areas. No added labor, just add it to cleaner, rinse or spray. Unlike most enzyme products, KOE is compatible with cleaners and germicides. You need to use only 1/4 oz. (not 1oz.) per gallon of water. Leaves a mild, fresh scent. Unpleasant animal-related odors are a common, daily occurrence, and managing them safely and effectively is an important part of keeping your work environment pleasant, your clients satisfied, and pets happy. Thornell specializes in creating products that permanently eliminate malodors - complex blends that work through counteraction, absorption, molecular bonding, inhibitors and residual actions.
Catalog No. 50-283-1081 Supplier Med Vet International Supplier No. KOE
Eliminates, does not mask, even old, impregnated odors from cages, runs, walls and other large areas. No added labor, just add it to cleaner, rinse or spray. Unlike most enzyme products, KOE is compatible with cleaners and germicides. You need to use only 1/4 oz. (not 1oz.) per gallon of water. Leaves a mild, fresh scent. Unpleasant animal-related odors are a common, daily occurrence, and managing them safely and effectively is an important part of keeping your work environment pleasant, your clients satisfied, and pets happy. Thornell specializes in creating products that permanently eliminate malodors - complex blends that work through counteraction, absorption, molecular bonding, inhibitors and residual actions.