Learn More
Koehler™ Instrument Ramsbottom Carbon Residue Apparatu: Control Bulbs
SDP
Catalog No. 1318710
Click to view available options
For Use With (Equipment):
Ramsbottom Carbon residue apparatus
Catalog No. For Use With (Equipment)
13-187-10 Ramsbottom Carbon residue apparatus
1 options
Catalog No. 13-187-10 Supplier Koehler™ Instrument Supplier No. K27200
Used with thermocouple pyrometer to verify compliance of furnace with ASTM performance requirements
Stainless-steel accessory for use with Ramsbottom Carbon Residue Apparatus.
Specifications
Type Control Bulb for Ramsbottom Carbon Residue Apparatus
For Use With (Equipment) Ramsbottom Carbon residue apparatus