Learn More
Cole-Parmer Koflo 3/8-21 316 SS Tube Mixer, 21 Elements; 3/8" OD
Catalog No. NC0895563 Shop All Cole-Parmer Products
Encompass_Preferred
These mixers consist of a series of fixed right- and left-hand elements. Tube and mixing elements are made of 316 stainless steel—ideal for mixing high-viscosity fluids such as epoxies. Mixers have plain ends and accept standard tube fittings
Catalog No. NC0895563 Supplier Cole-Parmer Supplier No. 0466962
These mixers consist of a series of fixed right- and left-hand elements. Tube and mixing elements are made of 316 stainless steel—ideal for mixing high-viscosity fluids such as epoxies. Mixers have plain ends and accept standard tube fittings