Learn More
Chemglass Life Sciences Drying Tower, KOH, Extra Coarse Frit, For Portable Vacuum System, Airfree, Schlenk
SDP
Catalog No. AF030050
Encompass_Preferred
Drying Tower, KOH, Extra Coarse Frit, For Portable Vacuum System, Airfree, Schlenk. Tower is designed to fit on cart system between the vacuum and pump oil trap. Trap has a sealed-in extra coarse fritted disc for support of KOH, and is 50mm ID x 250mm effective length with both ends having #20 o-ring joints. Lower adapter has a #20 o-ring joint with a 1" OD tubulation for connection to flexible vacuum tubing. Complete item consists of one KOH tower, one lower tubing adapter, one AF-0300-14, 90º connection adapter, two CG-150-06 #35 Stainless Steel pinch clamps and two #214 Viton o-rings. Approximately 18" of flexible tubing is required to connect the tower assembly to the AF-0300-08 oil pump trap along with two AF-0300-11 hose clamps. Tubing and clamps are NOT supplied with tower but can be ordered separately under AF-0300-12 and AF-0300-11.
Catalog No. AF030050 Supplier Chemglass Life Sciences Supplier No. AF030050
Drying Tower, KOH, Extra Coarse Frit, For Portable Vacuum System, Airfree, Schlenk. Tower is designed to fit on cart system between the vacuum and pump oil trap. Trap has a sealed-in extra coarse fritted disc for support of KOH, and is 50mm ID x 250mm effective length with both ends having #20 o-ring joints. Lower adapter has a #20 o-ring joint with a 1" OD tubulation for connection to flexible vacuum tubing. Complete item consists of one KOH tower, one lower tubing adapter, one AF-0300-14, 90º connection adapter, two CG-150-06 #35 Stainless Steel pinch clamps and two #214 Viton o-rings. Approximately 18" of flexible tubing is required to connect the tower assembly to the AF-0300-08 oil pump trap along with two AF-0300-11 hose clamps. Tubing and clamps are NOT supplied with tower but can be ordered separately under AF-0300-12 and AF-0300-11.