Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kojic acid, 100%, MP Biomedicals™

Catalog No. ICN15515701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15515701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15515701 Supplier MP Biomedicals, Inc Supplier No. 0215515701
Add to Cart
Add to Cart

Chemical Identifiers

CAS 501-30-4
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00006580
InChI Key BEJNERDRQOWKJM-UHFFFAOYSA-N
Synonym kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52
PubChem CID 3840
ChEBI CHEBI:43572
IUPAC Name 5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
SMILES OCC1=CC(=O)C(O)=CO1

Specifications

Color White
Assay Percent Range 1
Quantity 1 g
Formula Weight 142.1
Physical Form Crystalline Powder
Chemical Name or Material Kojic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.