Learn More
Selleck Chemical LLC Kojic acid S5174-25mg
SDP
Supplier:  Selleck Chemical LLC S517425MG
Encompass_Preferred
Kojic acid is a fungal metabolite that inhibits tyrosinase with an IC50 value of 30 6 M for mushroom tyrosinase It is a synthetic intermediate used for production of food additives
Catalog No. 50-311-1989