Learn More
Apexbio Technology LLC Kojic acid 501-30-4 200mg
SDP
Catalog No. 5000094543
Encompass_Preferred
Kojic acid (CAS 501-30-4) is a small-molecule inhibitor targeting tyrosinase a key enzyme involved in melanin biosynthesis It is designed to inhibit tyrosinase activity thereby reducing melanin production Kojic acid exerts its biological activity primarily through chelation of copper ions at the active site of tyrosinase In enzymatic assays on mushroom tyrosinase kojic acid demonstrates inhibitory activity with an IC50 typically ranging from 14 to 50 M Based on these pharmacological properties kojic acid holds research potential in hyperpigmentation disorders skin whitening applications and oxidative stress-related experimental models
Catalog No. 50-000-94543 Supplier Apexbio Technology LLC Supplier No. C7192200
Kojic acid (CAS 501-30-4) is a small-molecule inhibitor targeting tyrosinase a key enzyme involved in melanin biosynthesis It is designed to inhibit tyrosinase activity thereby reducing melanin production Kojic acid exerts its biological activity primarily through chelation of copper ions at the active site of tyrosinase In enzymatic assays on mushroom tyrosinase kojic acid demonstrates inhibitory activity with an IC50 typically ranging from 14 to 50 M Based on these pharmacological properties kojic acid holds research potential in hyperpigmentation disorders skin whitening applications and oxidative stress-related experimental models