Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kojic acid | 501-30-4 | MFCD00006580 | 100.0% | 142.11 g/mol | C6H6O4 | 1 ML
SDP

Catalog No. 5000210583
Encompass_Preferred

Kojic acid is a tyrosinase inhibitor supplied as a 10 mM solution in DMSO (1 mL) for research use. It inhibits melanin production by chelating copper at the tyrosinase active site and exhibits antioxidant, antimicrobial, and anti-inflammatory activities.

  • Concentration: 10 mM solution in DMSO.
  • Volume: 1 mL.
  • Cas number: 501-30-4.
  • Chemical formula: C6H6O4.
  • Molecular weight: 142.11 g/mol.
  • Purity: 99.99% (manufacturer stated).
  • Appearance: solution appearance not specified; solid described as off-white to light yellow.
  • Storage: in solvent, store at -80°C up to 6 months or -20°C up to 1 month.
  • Intended use: for research use only; not for human or patient use.

Catalog No. 50-002-10583 Supplier Medchemexpress LLC Supplier No. HYW05015410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Kojic acid is a tyrosinase inhibitor supplied as a 10 mM solution in DMSO (1 mL) for research use. It inhibits melanin production by chelating copper at the tyrosinase active site and exhibits antioxidant, antimicrobial, and anti-inflammatory activities.

  • Concentration: 10 mM solution in DMSO.
  • Volume: 1 mL.
  • Cas number: 501-30-4.
  • Chemical formula: C6H6O4.
  • Molecular weight: 142.11 g/mol.
  • Purity: 99.99% (manufacturer stated).
  • Appearance: solution appearance not specified; solid described as off-white to light yellow.
  • Storage: in solvent, store at -80°C up to 6 months or -20°C up to 1 month.
  • Intended use: for research use only; not for human or patient use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.