Learn More
Medchemexpress LLC Kojic acid | 501-30-4 | MFCD00006580 | 100.0% | 142.11 g/mol | C6H6O4 | 100 G
SDP
Catalog No. 5000209913
Encompass_Preferred
Kojic acid is a fungal metabolite used as a research reagent and reversible tyrosinase inhibitor for studies of melanin production, enzymology, and dermatological biology. It chelates copper at the tyrosinase active site and exhibits antioxidant and anti-inflammatory effects.
  • High purity analytical reagent (99.99%).
  • Suitable for tyrosinase inhibition assays and melanin research.
  • Available as a solid in multiple pack sizes up to 100 G.
  • Molecular formula C6H6O4, molecular weight 142.11 g/mol.
  • Manufacturer documents available: data sheet, SDS, and certificate of analysis for analytical use.
Catalog No. 50-002-09913 Supplier Medchemexpress LLC Supplier No. HYW050154100G
Kojic acid is a fungal metabolite used as a research reagent and reversible tyrosinase inhibitor for studies of melanin production, enzymology, and dermatological biology. It chelates copper at the tyrosinase active site and exhibits antioxidant and anti-inflammatory effects.
  • High purity analytical reagent (99.99%).
  • Suitable for tyrosinase inhibition assays and melanin research.
  • Available as a solid in multiple pack sizes up to 100 G.
  • Molecular formula C6H6O4, molecular weight 142.11 g/mol.
  • Manufacturer documents available: data sheet, SDS, and certificate of analysis for analytical use.