Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kojic Acid Sodium Salt Hydrate 98.0+%, TCI America™
SDP

Catalog No. K00365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K00365G Supplier TCI America Supplier No. K00365G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70145-54-9
Molecular Formula C6H7NaO5
Molecular Weight (g/mol) 182.107
MDL Number MFCD00191537
InChI Key BENOTXBUMHIXTM-UHFFFAOYSA-N
Synonym 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one Sodium Salt
PubChem CID 91658936
IUPAC Name sodium;(5-hydroxy-4-oxopyran-2-yl)methanolate;hydrate
SMILES C1=C(OC=C(C1=O)O)C[O-].O.[Na+]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 164.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Kojic Acid Sodium Salt Hydrate
RTECSNumber UQ0950000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.