Learn More
Med Vet International Jorvet Kitty and Kanine Kollar, Medium, J1085B
Catalog No. 5000495690
Encompass_Preferred
The Jorvet medium Petitie Kitty and Kanine Kollar was designed to be a sensible solution to the everyday patient management of an esophagostomy tube in cats and dogs. This collar has a neck size of 12-16in., is adjustable to fit securely around the animals neck, with a hole for the tube and a hook and loop strap to secure it. Circular cotton pads are supplied to place between the collar and the skin to protect the wound and absorb drainage from below. This collar for cats and dogs helps eliminate the need for owners to attempt to learn bandaging tecniques, as well as unnecessary repeated visits to your veterinary office for re-bandaging. Features: Machine washable collar Ease of application and removal Lowered risk of tube removal Decreased risk of wound trauma Can be left in place for Radiographs, MRI, Chemotherapy, or other procedures Aesthetically pleasing for squemish clients
Catalog No. 50-004-95690 Supplier Med Vet International Supplier No. JOR1085B
The Jorvet medium Petitie Kitty and Kanine Kollar was designed to be a sensible solution to the everyday patient management of an esophagostomy tube in cats and dogs. This collar has a neck size of 12-16in., is adjustable to fit securely around the animals neck, with a hole for the tube and a hook and loop strap to secure it. Circular cotton pads are supplied to place between the collar and the skin to protect the wound and absorb drainage from below. This collar for cats and dogs helps eliminate the need for owners to attempt to learn bandaging tecniques, as well as unnecessary repeated visits to your veterinary office for re-bandaging. Features: Machine washable collar Ease of application and removal Lowered risk of tube removal Decreased risk of wound trauma Can be left in place for Radiographs, MRI, Chemotherapy, or other procedures Aesthetically pleasing for squemish clients