Learn More
Sigma Aldrich Fine Chemicals Biosciences Kolliphor® P 188, 9003-11-6, MFCD00082049, 1 kg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 157591KG
Encompass
Synonym: Lutrol® F68, Poloxamer 188, Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol). Kolliphor® P 188 has been widely used as a thickening agent and gel binder. This research grade product is intended for use in R&D and development only. Kolliphor® is a registered trademark of BASF SE.
Catalog No. NC0917244