Learn More
ATCC Gluconacetobacter xylinus
Catalog No. 502383435
Encompass
Komagataeibacter oboediens strain [NCIB 8034, NRC 6018] is a whole-genome sequenced bacterium that produces 2-keto-L-gulonic acid (provitamin C) by enzymatic conversion of sorbitol.
Catalog No. 50-238-3435 Supplier ATCC Supplier No. 10245
 Restricted Item
Add to Cart
Add to Cart
Komagataeibacter oboediens strain [NCIB 8034, NRC 6018] is a whole-genome sequenced bacterium that produces 2-keto-L-gulonic acid (provitamin C) by enzymatic conversion of sorbitol.