Learn More
ATCC Enterococcus faecalis; Strain: YIT0072
Catalog No. 502379624
Encompass
Probiotics production. Sensitive to ampicillin, vancomycin, erythromycin, and tetracycline.
Catalog No. 50-237-9624 Supplier ATCC Supplier No. BAA2820
Probiotics production. Sensitive to ampicillin, vancomycin, erythromycin, and tetracycline.