Learn More
MSC Komelon Measuring Wheel, 14.3 in Diameter, 10,000 ft Measurement, Pistol Grip Handle
Catalog No. 5000528349
Encompass
This measuring wheel is designed for accurate outdoor distance measurements. It features a durable 14.3 inch diameter wheel and a gear-driven counter that reads up to 10,000 feet, making it suitable for various professional applications. The telescoping handle with a comfortable pistol grip ensures ease of use and portability, while the composite tread provides improved traction on diverse surfaces.
  • 14.3 inch diameter wheel
  • Gear driven counter with push button reset
  • Telescoping handle with comfortable pistol grip
  • Five digit counter reads up to 10,000 feet
  • Durable for outdoor use
  • Composite tread for improved traction on smoother surfaces
  • Protective rubber counter jacket
  • Spring loaded kickstand
Catalog No. 50-005-28349 Supplier MSC Supplier No. 32514648
This measuring wheel is designed for accurate outdoor distance measurements. It features a durable 14.3 inch diameter wheel and a gear-driven counter that reads up to 10,000 feet, making it suitable for various professional applications. The telescoping handle with a comfortable pistol grip ensures ease of use and portability, while the composite tread provides improved traction on diverse surfaces.
  • 14.3 inch diameter wheel
  • Gear driven counter with push button reset
  • Telescoping handle with comfortable pistol grip
  • Five digit counter reads up to 10,000 feet
  • Durable for outdoor use
  • Composite tread for improved traction on smoother surfaces
  • Protective rubber counter jacket
  • Spring loaded kickstand