Learn More
Beckman Coulter TUBES PP KON 1 X 3-1/2 (PKG-50)
Catalog No. NC9885042
Encompass_Preferred
Konical Tube, Standard, Polypropylene, 31.5mL, 25x89mm, Quantity: 50/Box
Catalog No. NC9885042 Supplier Beckman Coulter Supplier No. 358126
Konical Tube, Standard, Polypropylene, 31.5mL, 25x89mm, Quantity: 50/Box