Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Amber 203 Serum Vials
SDP

Catalog No. p-313434 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
2 mL
10 mL
30 mL
Height (Metric):
40 mm
54 mm
86 mm
Diameter (Metric) Outer:
15 mm
25 mm
30 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Height (Metric) Diameter (Metric) Outer
10-310-243C 10 mL 54 mm 25 mm
22-268786 2 mL 40 mm 15 mm
10-310-243E 30 mL 86 mm 30 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-310-243C Supplier DWK Life Sciences Supplier No. 62421D10
Only null left
Add to Cart
Add to Cart

Autoclavable; ideally suited for the handling, and storage of a variety of liquids including reagents, vaccines, blood plasma, culture media, chromatography.

  • Serum vial with an aluminum seal finish
  • Comes without a closure and is lighter in weight than molded bottles
  • Manufactured from amber 203 borosilicate glass
  • Enhanced by the most popular GPI aluminum seal finishes, this line offers a variety of choices

TRUSTED_SUSTAINABILITY

Specifications

Color Amber
Diameter (Metric) Outer 25 mm
Material Borosilicate Glass
Volume (Metric) 10 mL
For Use With (Application) Ideally suited for the handling, and storage of a variety of liquids including reagents, vaccines, blood plasma, culture media, chromatography samples
Material (Seal) Aluminum
Volume (English) 2.75 dr.
Autoclavable Autoclavable
Certifications/Compliance ASTM E438, Type I
Height (Metric) 54 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.