Learn More
DWK Life Sciences KIMBLE™ KONTES™ Bevel-Seal™ PTFE Inlet Adapter
SDP
Catalog No. K1798002224 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No.:
24/25
Catalog No. Standard Taper Joint No.
K179800-2224 24/25
1 options
Catalog No. K179800-2224 Supplier DWK Life Sciences Supplier No. 1798002224
Add to Cart
Add to Cart
Includes
Adapter, cap and Viton™ O-ring
KIMBLE™ KONTES™ Bevel-Seal™ PTFE Inlet Adapter provides a vacuum-tight seal using a compression cap and O-ring, allowing adjustable immersion of thermometers and similar instruments. The cap is rated for use up to 150°C (302°F).
For use with pipets, syringes, or other small items with outer diameters from 0.06 to 0.62 in. (2 to 17mm)
  • Provides a vacuum-tight seal with compression cap and O-ring to allow adjustable immersion of thermometers and similar instruments
  • Cap can be used up to 150°C (302°F)
  • Made from PTFE for excellent chemical resistance
  • Bevel-Seal design ensures a secure, reliable connection
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For use with pipets, syringes, or other small items with outer diameters from 0.06 to 0.62 in.
Includes Adapter, cap and Viton™ O-ring
Material Borosilicate Glass
Product Type Bevel-Seal™ Inlet Adapter
Standard Taper Joint No. 24/25