Learn More
DWK Life Sciences Kimble™ Kontes™ Arsine Generator Scrubber
SDP
Catalog No. K2411012440 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
24/40, 12/2
Catalog No. Standard Taper Joint
K2411012440 24/40, 12/2
1 options
Catalog No. K2411012440 Supplier DWK Life Sciences Supplier No. 2411012440
Add to Cart
Add to Cart
This Arsine Generator Scrubber is a Component Part For Arsine Generators
  • Arsine Generator Scrubber Only
  • Standard Taper 24/4
  • Spherical Joint 12/2
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Product Type Scrubber
Standard Taper Joint 24/40, 12/2
For Use With (Equipment) For arsine generator