Learn More
DWK Life Sciences KIMBLE™ KONTES™ Component Parts for MIDI-VAP 4000 Ammonia and Phenol Distillation System
Shop All DWK Life Sciences Products
Description
- Replacement and spare component parts for the Kimble™ Kontes™ MIDI-VAP 4000 Ammonia and Phenol Distillation System
- Constructed from borosilicate glass for chemical resistance and durability
- Designed for use in multi-position distillation workflows for ammonia and phenol analysis
- Compatible with the MIDI-VAP 4000 10-position manifold heater system
- Sold individually to allow targeted replacement of specific glassware components
Specifications
Specifications
| Type | Distillation Apparatus |
| For Use With (Equipment) | MIDI-VAP™ 3000 ammonia distillation apparatus |