Learn More
DWK Life Sciences™ Kimble™ Kontes™ 4.5 in. Distilling Head for Ammonia/Phenol Midi-Vap™ 4000
SDP
Catalog No. K4791564501 Shop All DWK Life Sciences Products
Change view
Click to view available options
Type:
4.5 in. Distilling Head
Catalog No. Type
K4791564501 4.5 in. Distilling Head
1 options
Catalog No. K4791564501 Supplier DWK Life Sciences™ Supplier No. 4791564501
Add to Cart
Add to Cart
Replacement component: 4.5 in. distilling head for Ammonia/Phenol MIDI-VAP™ 4000
  • Glassware manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Type 4.5 in. Distilling Head
For Use With (Equipment) MIDI-VAP™ 3000 ammonia distillation apparatus