Learn More
DWK Life Sciences™ Kimble™ Kontes™ Flask Inlet Adapter, Monier-Williams, 24/40 Joint
SDP
Catalog No. K5138010000 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
24/40
Catalog No. Standard Taper Joint
K513801-0000 24/40
1 options
Catalog No. K513801-0000 Supplier DWK Life Sciences™ Supplier No. 5138010000
Add to Cart
Add to Cart
Component Parts of Monier-Williams Sulfites Distillation Apparatus which is used for the determination of free sulfite and a reproducible portion of the bound sulfites in food, such as the carbonyl addition products.
  • Flask Inlet Adapter
  • Standard Taper joint 24/40
  • Case of 1
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Type Flask Inlet Adapter
Standard Taper Joint 24/40