Learn More
DWK LIFE SCIENCES™ KIMBLE™ KONTES™ Distillation Head for Microflex Kit
SDP
Catalog No. K7460800000 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
For microflex kit
Catalog No. For Use With (Equipment)
K746080-0000 For microflex kit
1 options
Catalog No. K746080-0000 Supplier DWK LIFE SCIENCES™ Supplier No. 7460800000
KIMBLE™ KONTES™ Distillation Head for Microflex Kit is a Hickman-style distillation head designed for use with the KONTES™ Microflex distillation kit, enabling small-scale and microscale distillations in the laboratory.
  • Hickman-style distillation head designed for use with the Kontes Microflex microscale distillation kit
  • Enables small-scale and microscale distillations
  • Manufactured by Kimble™ Kontes™, a DWK Life Sciences brand
  • Designed for compatibility with Microflex kit components
TRUSTED_SUSTAINABILITY
Specifications
Type Hickman Distillation Head
For Use With (Equipment) For microflex kit