Learn More
DWK Life Sciences Kimble™ Kontes™ Connecting Distilling Adapter Sub-boiling
SDP
Catalog No. K1695002440 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No.:
24/40
Catalog No. Standard Taper Joint No.
K169500-2440 24/40
1 options
Catalog No. K169500-2440 Supplier DWK Life Sciences Supplier No. 1695002440
Add to Cart
Add to Cart
Useful in the sub-boiling separation of mixtures having volatile components.
  • Standard Taper inner joints are parallel
  • Joints are the same size
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Application) Useful in the sub-boiling separation of mixtures having volatile components
Material Borosilicate Glass
Product Type Connecting Distillation Adapter
Quantity 1
Standard Taper Joint No. 24/40