Learn More
DWK Life Sciences Kimble™ Kontes™ Four-Neck Round-Bottom Heavy-Wall Flasks with Vertical Necks
SDP
Catalog No. p-3247061 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
3000 mL
5 L
Standard Taper Joint:
29/42
29/42 Center, 24/42 Side
34/45 Center, 24/40 Side
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Standard Taper Joint
K6070000724 1000 mL 29/42 Center, 24/42 Side
K607000-1529 3000 mL 29/42
K607000-2024 5 L 34/45 Center, 24/40 Side
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K6070000724 Supplier DWK Life Sciences Supplier No. 6070000724
Only null left
Add to Cart
Add to Cart
With standard taper outer joints and straight side necks
  • Heavy wall
  • Four neck round-bottom flask
  • Outer joints all vertical
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Neck Style Vertical
Standard Taper Joint 29/42 Center, 24/42 Side