Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ Kontes™ Kem-Kit™ Semi-Macro 19/22 Organic Chemistry Kit
SDPSCI_ED

Catalog No. S65959 Shop All DWK Life Sciences Products
Change view
Click to view available options
Kem Kit Semi-macro 19/22
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S65959 Kem Kit Semi-macro 19/22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65959 Supplier Kimble™ Supplier No. 2698000000/EMD

missing translation for 'validMainframeMsg'

Compact, 14-piece kit for organic chemistry studies

  • Selection of basic small-scale glassware with standard taper 19/22 joints
  • Performs extraction, reflux, reflux with addition, fractionation, distillation (steam, atmospheric, and vacuum), and other organic chemistry applications
  • Supplied in a heavy-duty, double-wall polyethylene storage case with handle and hinged lid
  • Case dimensions: 13.5L × 11W × 5.5 in.H (34 × 28 × 14cm)

Order Info

  • Comprehensive instruction manual
  • Storage case

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
For Use With (Application) For extraction, reflux, reflux with addition, fractionation, and distillation
Includes Bleed adapter, claisen adapter, condenser, connecting adapter, distillation column, 25mL, 50mL, 100mL, 250mL, 500mL Flask, 125mL PTFE funnel, outlet adapter, stopper, vacuum adapter
Standard Taper Joint 19/22
Product Type Standard Taper Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.