Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ KONTES™ KEM-KLAMP Solid Pennyhead Glass Stopper with Lug
SDP

Catalog No. K8518002440 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Borosilicate Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
K851800-2440 Borosilicate Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K851800-2440 Supplier DWK Life Sciences Supplier No. 8518002440
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The KIMBLE™ KONTES™ KEM-KLAMP Solid Pennyhead Glass Stopper with Lug is designed for use with 675500 KEM-KLAMPS™. Features a pennyhead top with lug attachment and is manufactured from low extractable borosilicate glass meeting USP Type I and ASTM E438 requirements.

  • Pennyhead top has a lug attachment to engage the spring portion of the KEM-KLAMP™
  • Designed for use with 675500 KEM-KLAMPS™
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY

Specifications

Shape Tapered
Material Borosilicate Glass
Autoclavable Autoclavable
Color Clear
Description Includes: Lug
For Use With (Equipment) Standard taper kem-klamp
Standard Taper Joint 24/40
Product Type Pennyhead Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.