Learn More
DWK Life Sciences KIMBLE™ KONTES™ Neoprene Gasket for NMR Tube Washer
SDP
Catalog No. K4599510000 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Application):
For NMR utility washer
Catalog No. For Use With (Application)
K4599510000 For NMR utility washer
1 options
Catalog No. K4599510000 Supplier DWK Life Sciences Supplier No. 4599510000
Add to Cart
Add to Cart
KIMBLE™ KONTES™ Neoprene Gasket for NMR Tube Washer is a replacement gasket compatible with KONTES™ utility washers designed for rapid, safe washing and drying of cuvettes, tubes, and small-scale volumetric items.
  • Replacement neoprene gasket for Kimble Kontes NMR Tube Washer utility washers
  • Utility washers are used for rapid, safe washing and drying of cuvettes, tubes, and small-scale volumetric items
  • Unit is supplied with one gasket, a tube washer, an Erlenmeyer flask, and a 3/8 in. hose connection
  • Replacement gaskets sold as Case of 12
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For NMR utility washer
Material Neoprene
Type Gasket