Learn More
KIMBLE™ KONTES™ Pear Shaped Boiling Flask, 19/22
SDP
Catalog No. 01672728 Shop All DWK Life Sciences Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
01-672-728 Autoclavable
1 options
Catalog No. 01-672-728 Supplier KIMBLE™ Supplier No. 2942600015
Boiling flask manufactured from 33 expansion, low extractable borosilicate glass.
  • Heavy wall
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 15 mL
Material Borosilicate Glass
Product Type Boiling Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Sterility Non-sterile
Bottom Shape Round