Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ KONTES™ Medium Length Pennyhead Stoppers, 14/20
SDP

Catalog No. K8505001420 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Borosilicate Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
K850500-1420 Borosilicate Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K850500-1420 Supplier DWK Life Sciences Supplier No. 8505001420
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KIMBLE™ KONTES™ Medium Length Pennyhead Stoppers, 14/20. KIMBLE™ KONTES™ Medium Length Pennyhead Stoppers are used with laboratory glassware such as flasks, mixing cylinders and separatory funnels. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E

  • Sizes Standard Taper 10/18 through Standard Taper 19/22 are solid
  • Manufactured from 33 expansion, low extractable borosilicate glass, not hollow
  • Conforms to USP Type I and ASTM E438, Type I, Class A requirements
  • Designed for use with laboratory glassware including flasks, mixing cylinders and separatory funnels
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Material Borosilicate Glass
Shape Tapered
Color Clear
Standard Taper Joint 14/20
For Use With (Equipment) Laboratory glassware such as flasks and separatory funnels
Product Type Pennyhead Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.