Learn More
DWK Life Sciences Kimble™ Kontes™ Full Length Pennyhead Stoppers, 10/30
SDP
Catalog No. K8510001030 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Glass
Catalog No. Material
K851000-1030 Glass
1 options
Catalog No. K851000-1030 Supplier DWK Life Sciences Supplier No. 8510001030
Add to Cart
Add to Cart
Stoppers are used with laboratory glassware such as flasks, mixing cylinders and separatory funnels.
  • Sizes Standard Taper 10/30 through Standard Taper 19/38 are solid
  • Manufactured from 33 expansion, low extractable borosilicate glass - Not hollow
  • Conforms to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
For Use With (Equipment) Laboratory glassware such as flasks and separatory funnels
Standard Taper Joint 10/30
Product Type Pennyhead Stopper