Learn More
DWK Life Sciences Kimble™ Kontes™ Polyethylene Stoppers
SDP
Catalog No. K7742400014 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
14/10, 14/20, 14/35
19/22 ,19/38
24/35, 24/40
29/26, 29/42
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
K7742400014 14/10, 14/20, 14/35
K7742400019 19/22 ,19/38
K7742400024 24/35, 24/40
K7742400029 29/26, 29/42
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. K7742400014 Supplier DWK Life Sciences Supplier No. 7742400014
Add to Cart
Add to Cart
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
  • Leak-proof, air-tight seals
  • Wide top flange for ease of use
TRUSTED_SUSTAINABILITY
Specifications
Product Type Plug Stopper
Material Polyethylene
Standard Taper Joint 14/10, 14/20, 14/35