Learn More
Kimble™ Kontes™ PTFE-Plug Stopcocks, Pressure Equalizing Lines, Ground Joints, standard taper Stopper Funnels
SDP
Catalog No. K2992300050 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
50 mL
Catalog No. Capacity (Metric)
K299230-0050 50 mL
1 options
Catalog No. K299230-0050 Supplier Kimble™ Supplier No. 2992300050
  • Ground top and bottom joints
  • Pressure equalizing lines permit addition of material into a closed system
  • Drip tips extend below the standard taper joints to ensure delivery
  • Glass stoppers have hexagonal heads for easy gripping
  • Set stoppers down on their flattened tops to avoid contaminating ground surface
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Material Glass