Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ Kontes™ Reactor Bottom Outlet Flush Seal PTFE Plug Valves
SDP

Catalog No. p-3246490 Shop All DWK Life Sciences Products
Change view
Click to view available options
:
Reactor Bottom Outlet, Valve Body, 1 in. BP
Reactor Bottom Outlet, Valve Body, 2 in. BP
Reactor Bottom Outlet, Valve Plug, 1 in. BP
Reactor Bottom Outlet, Valve Plug, 2 in. BP
Reactor Bottom Outlet, Valve Seat, 1 in. BP
Reactor Bottom Outlet, Valve Seat, 2 in. BP
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
K607731-0010 Reactor Bottom Outlet, Valve Plug, 1 in. BP
K607731-0020 Reactor Bottom Outlet, Valve Plug, 2 in. BP
K607732-0010 Reactor Bottom Outlet, Valve Body, 1 in. BP
K607732-0020 Reactor Bottom Outlet, Valve Body, 2 in. BP
K607733-0010 Reactor Bottom Outlet, Valve Seat, 1 in. BP
K607733-0020 Reactor Bottom Outlet, Valve Seat, 2 in. BP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. K607731-0010 Supplier Kimble™ Supplier No. 6077310010
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.