Learn More
DWK Life Sciences Kimble™ Kontes™ Enlarging Connecting Adapters 14/20 to 24/40
SDP
Catalog No. K2747500242 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No.:
24/40 Inner Bottom, 14/20 Outer Top
Catalog No. Standard Taper Joint No.
K274750-0242 24/40 Inner Bottom, 14/20 Outer Top
1 options
This item is not returnable. View return policy
Catalog No. K274750-0242 Supplier DWK Life Sciences Supplier No. 2747500242
Add to Cart
Add to Cart
This item is not returnable. View return policy
For enlarging the size of Standard Taper glass joints from 14/20 outer to 24/40 inner.
  • Outer joint is at the top, inner at the bottom
  • Top (outer) joint 14/20, Bottom (inner) joint 24/40
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Application) For enlarging the size of Standard Taper glass joints
Material Borosilicate Glass
Product Type Enlarging Connecting Adapter
Standard Taper Joint No. 24/40 Inner Bottom, 14/20 Outer Top