Learn More
DWK Life Sciences KIMBLE™ KONTES™ TLC Plate Streaker Reservoir Only
SDP
Catalog No. K4164320000 Shop All DWK Life Sciences Products
Catalog No. K416432-0000 Supplier DWK Life Sciences Supplier No. 4164320000
Add to Cart
Add to Cart
The KIMBLE™ KONTES™ TLC Plate Streaker Reservoir Only is a replacement glass reservoir for the KIMBLE™ KONTES™ TLC Plate Streaker. Made from 33-expansion, low-extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Replacement Reservoir for use with Kimble™ 413430-0000.
  • Replacement glass reservoir for the Kimble™ Kontes™ TLC Plate Streaker (416430-0000)
  • Fits the adjustable holder attached to the stainless steel base assembly
  • Manufactured from 33-expansion, low-extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Reservoir is filled with the solution to be chromatographed; moving the unit back and forth along the counter edge produces a uniform streak on the TLC plate
  • Sold individually as a replacement part; compatible with base assembly (416431-0000), glass capillary set (416433-0000), and stainless steel capillary set (416434-0000)
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Quantity 1 Ea.
Type Reservoir
For Use With (Equipment) TLC Chromatography Plate Streaker for Uniformly Streaking TLC Plates