Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ KIMBLE™ TLC Plate Streaker Glass Capillary Set Only
SDP

Catalog No. K4164330000 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
K416433-0000 Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K416433-0000 Supplier DWK LIFE SCIENCES™ Supplier No. 4164330000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement glass capillary set for use with KIMBLE™ TLC Plate Streakers

Replacement Glass Capillary Set for use with Kimble™ 413430-0000.

  • Includes 2 small and 2 large glass capillaries
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • TLC Plate Streaker is a simple device for uniformly streaking TLC plates
  • Streaker features a stainless steel base with an adjustable holder for a glass reservoir
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP Type I and ASTM E438 Type I Class A
Material Glass
Sterility Non-sterile
For Use With (Application) Uniformly Streaking TLC Plates
Includes 2 Small and 2 Large Glass Capillaries
Quantity 1 Set
Color Clear
Type Capillary Set
Disposable Single-use
For Use With (Equipment) TLC Chromatography Plate Streaker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.