Learn More
DWK Life Sciences KIMBLE™ TLC Plate Streaker Stainless Steel Capillary Set Only
Shop All DWK Life Sciences Products
Description
- Replacement stainless steel capillary set for use with Kimble™ TLC Plate Streaker (413430-0000)
- Includes one small and one large stainless steel capillary
- Compatible with the Kimble™ TLC Plate Streaker base assembly for uniform streaking of TLC plates
- TLC Plate Streaker is a simple device for uniformly streaking TLC plates using an adjustable holder and capillary reservoir
Specifications
Specifications
| Material | Stainless Steel |
| Quantity | 1 Ea. |
| Type | Stainless Steel Capillary Set |
| For Use With (Equipment) | TLC Chromatography Plate Streaker for Uniformly Streaking TLC Plates |
| Includes | 1 Small and 1 Large Stainless Steel capillaries |