Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Full Length Stoppers
SDP
Catalog No. K8510003445 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
29/42
34/45
45/50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
K851000-3445 34/45
K851000-2942 29/42
K851000-4550 45/50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K851000-3445 Supplier DWK Life Sciences Supplier No. 8510003445
Add to Cart
Add to Cart
Stoppers are used with laboratory glassware such as flasks and separatory funnels.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Flask and separatory funnel
Standard Taper Joint 34/45
Material Glass
Product Type Solid Pennyhead Stopper