Learn More
DWK Life Sciences Kimble™ Kontes™ Stirrer Assembly with PTFE Tri-Blade on PTFE-Shielded Stainless-Steel Shaft
SDP
Catalog No. K7850500629 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
Joint: 29/42
Joint: 45/50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
K7850500629 Joint: 29/42
K7850500645 Joint: 45/50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K7850500629 Supplier DWK Life Sciences Supplier No. 7850500629
Complete unit includes a Hi-Vac™ PTFE and glass stirrer bearing and PTFE-shielded stir shaft with PTFE tri-blade. This assembly is recommended for reaction systems requiring vacuums which have 4 in. or larger flange.
TRUSTED_SUSTAINABILITY
Specifications
Standard Taper Joint Joint: 29/42