Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ KONTES™ Storage Caps for HPLC Ultra-Ware Plastic-Coated Conical Bottom Reservoirs
SDP

Catalog No. K9539080000 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Application):
For HPLC Ultra-Ware™ plastic-coated conical bottom reservoirs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
K953908-0000 For HPLC Ultra-Ware™ plastic-coated conical bottom reservoirs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K953908-0000 Supplier DWK Life Sciences Supplier No. 9539080000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KIMBLE™ KONTES™ Storage Caps for HPLC Ultra-Ware Plastic-Coated Conical Bottom Reservoirs provide a secure seal for GL 45 threaded conical bottom reservoirs used in liquid chromatography mobile phase preparation, storage, and delivery.

  • Designed for use with Kimble™ Kontes™ ULTRA-WARE Plastic-Coated Conical Bottom HPLC Reservoirs
  • Compatible with GL 45 screw thread reservoirs
  • PTFE construction for broad chemical resistance suitable for HPLC mobile phase solvents
  • 45 mm cap size fits standard ULTRA-WARE GL 45 reservoir openings
  • Supports safe storage and sealing of liquid chromatography mobile phases
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
For Use With (Application) For HPLC Ultra-Ware™ plastic-coated conical bottom reservoirs
Includes GL45 screw thread
Material PTFE
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Reusable
Product Type Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.