Learn More
DWK Life Sciences Kimble™ Kontes™ Straight Thermometer Inlet Adapters
SDP
Catalog No. K1970002440 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No.:
24/40 Bottom, 24/40 Top
Catalog No. Standard Taper Joint No.
K197000-2440 24/40 Bottom, 24/40 Top
1 options
Catalog No. K197000-2440 Supplier DWK Life Sciences Supplier No. 1970002440
Add to Cart
Add to Cart
With hose connection on side
  • 24/40 joints bottom
  • 24/40 joints top
  • No. 2 hose connection
  • 155mm height
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Fits Tubing ID (English) 0.37 in.
Fits Tubing ID (Metric) 9.39mm
Height (English) 6.1 in.
Height (Metric) 155mm
Includes Side hose connection no. 2
Material Borosilicate Glass
Product Type Straight Thermometer Inlet Adapter
Standard Taper Joint No. 24/40 Bottom, 24/40 Top