Description
- Replacement base assembly for use with Kimble™ TLC Plate Streaker (416430-0000)
- Stainless steel base with an adjustable holder for a glass reservoir
- Glassware manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
- Streaker is placed over the edge of the counter top; plate to be streaked is positioned parallel to it
- Bent capillary tip is adjusted to lightly touch the plate; reservoir is filled with the solution to be chromatographed
- Moving the unit back and forth along the counter edge produces a uniform streak
Specifications
Specifications
| Material | Stainless Steel |
| Quantity | 1 Ea. |
| Type | TLC Plate Streaker Base |
| For Use With (Equipment) | TLC Chromatography Plate Streaker |
| Includes | Adjustable holder for a Glass reservoir |