Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Vacuum Filtration Adapters
SDP

Catalog No. p-2829983 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
24/40
For Use With (Equipment):
Fits No. 2 pluro stopper
Fits No. 3 pluro stopper
Fits No. 4 pluro stopper
No. 5 and 6 Pluro Stopper
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No. For Use With (Equipment)
K178110-2440 24/40 No. 5 and 6 Pluro Stopper
K17812-00014 14/20 Fits No. 2 pluro stopper
K17812-00024 24/40 Fits No. 3 pluro stopper
K17812-00124 24/40 Fits No. 4 pluro stopper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. K178110-2440 Supplier DWK Life Sciences Supplier No. 1781102440
Only null left
Add to Cart
Add to Cart

Use for reduced pressure filtration with plain stem Buchner funnels.

  • Top designed with a flange to accommodate a pluro stopper
  • Pluro stopper sold separately
  • Includes a lower drip tip
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Application) Use for reduced pressure filtration with plain stem buchner funnels
Fits Tubing ID (English) 0.37 in.
Fits Tubing ID (Metric) 9.39mm
For Use With (Equipment) No. 5 and 6 Pluro Stopper
Includes Hose connection
Material Borosilicate Glass
Product Type Vacuum Filtration Adapter
Standard Taper Joint No. 24/40
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.