Learn More
Kimble™ Kontes™ Volatile Organic Sampling Train Trap
SDP
Catalog No. K5911800000 Shop All DWK Life Sciences Products
Change view
Click to view available options
VOST Trap
Catalog No.
K591180-0000 VOST Trap
1 options
Catalog No. K591180-0000 Supplier Kimble™ Supplier No. 5911800000
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For air sampling. Use VOST (Volatile Organic Sampling Train) trap to sample and analyze hazardous waste incinerator stack emissions to verify the destruction and removal efficiencies of volatile organics.