Learn More
Sigma Aldrich Fine Chemicals Biosciences KontesR NMR tubes 5 mm dia
Supplier:  Sigma Aldrich Fine Chemicals Biosciences KN8971937100100EA
Encompass
5 mm disposable tubes are manufactured from N-51A borosilicate glass. Wall thickness for these tubes is 0.015 in. Supplied without a sandblasted marking spot in a bundled multi-pack.Caps are included but not attached.
Catalog No. 50-290-0717