Learn More
Sigma Aldrich Fine Chemicals Biosciences KontesR NMR tubes 5 mm dia
Supplier:  Sigma Aldrich Fine Chemicals Biosciences KN89723000005EA
Encompass
Each and every precision 5 mm tube is 100% gauged to meet the most exacting standards. These tubes are made of KG-33 borosilicate glass and are ideal for sealing directly to vacuum manifolds joints or valves. Wall thickness for these precision tubes is 0.38 mm.Caps are attached. All tubes supplied in packs of 5.
Catalog No. 50-290-0702