Learn More
Sigma Aldrich Fine Chemicals Biosciences KontesR NMR tubes 5 mm dia
Catalog No. 502900086
Encompass
5 mm thrift tubes are manufactured from N-51A borosilicate glass. Wall thickness for these tubes is 0.38 mm.Caps are attached. All tubes supplied in packs of 5.
Catalog No. 50-290-0086 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. KN89714000075EA
5 mm thrift tubes are manufactured from N-51A borosilicate glass. Wall thickness for these tubes is 0.38 mm.Caps are attached. All tubes supplied in packs of 5.